Skip to Content
Merck
All Photos(1)

Key Documents

W419501

Sigma-Aldrich

Phthalide

98%

Synonym(s):

1-Isobenzofuranone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O2
CAS Number:
Molecular Weight:
134.13
FEMA Number:
4195
Beilstein:
114632
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.056

biological source

synthetic

Assay

98%

bp

290 °C (lit.)

mp

71-74 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

coconut

SMILES string

O=C1OCc2ccccc12

InChI

1S/C8H6O2/c9-8-7-4-2-1-3-6(7)5-10-8/h1-4H,5H2

InChI key

WNZQDUSMALZDQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Song-Hwa Chae et al.
Journal of agricultural and food chemistry, 59(15), 8193-8198 (2011-07-07)
The residual contact toxicity of three benzofuranoids (Z)-butylidenephthalide (1), (3S)-butylphthalide (2), and (Z)-ligustilide (3) identified in the rhizome of Cnidium officinale (Apiaceae) to B- and Q-biotype females of Bemisia tabaci was evaluated using a leaf-dip bioassay. Results were compared with
Chang-Yin Li et al.
Journal of pharmaceutical and biomedical analysis, 55(1), 146-160 (2011-02-01)
In this work, the metabolite profiles of Danggui Buxue Tang (DBT) in rat bile and plasma were qualitatively described, and the possible metabolic pathways of DBT were subsequently proposed. Emphasis was put on correlative analysis of metabolite profiling in different
Jie Luo et al.
Chemical communications (Cambridge, England), 48(39), 4707-4709 (2012-04-11)
The first Mannich reaction employing phthalides using a quinidine-based multifunctional catalyst has been developed. The reported method led to the synthesis of 3,3-disubstituted phthalide derivatives in excellent yields, with good diastereo- and enantioselectivities. Convenient synthesis of chiral isoquinolinones and isoquinolines
Bin Xiao et al.
Bioorganic & medicinal chemistry, 20(16), 4954-4961 (2012-07-24)
On the basis of a marine fungal phthalide (paecilocin A) skeleton, we synthesized 20 analogs and evaluated them for peroxisome proliferator-activated receptor gamma (PPAR-γ) binding and activation. Among these analogs, 6 and 7 had significant PPAR-γ binding activity, and 7
Yun Wei et al.
Journal of chromatography. A, 1284, 53-58 (2013-03-15)
The phthalide compounds of Chuanxiong rhizoma including senkyunolide A, levistolide A, Z-ligustilide and 3-butylidenephthalide, have been reported as the biologically active compounds because of their therapeutic effects. In this work, online high-speed counter-current chromatography coupled with semi-preparative liquid chromatography instrument

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service