Skip to Content
Merck
All Photos(1)

Key Documents

W415901

Sigma-Aldrich

2-Methoxythiophenol

≥97%, FG

Synonym(s):

2-Mercaptoanisole, 2-Methoxybenzenethiol, Thioguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4SH
CAS Number:
Molecular Weight:
140.20
FEMA Number:
4159
Beilstein:
2042178
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.139
NACRES:
NA.21
Organoleptic:
meaty; spicy; smoky
grade:
FG
Halal
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.591 (lit.)

bp

99 °C/8 mmHg (lit.)

solubility

H2O: insoluble
alcohol: soluble

density

1.152 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; spicy; smoky

SMILES string

COc1ccccc1S

InChI

1S/C7H8OS/c1-8-6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

DSCJETUEDFKYGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Methoxythiophenol is a sulfur-containing compound that can be used as a flavoring agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acute toxicity of thioguaiacol and of versalide in rodents.
K R Butterworth et al.
Food and cosmetics toxicology, 19(6), 753-755 (1981-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service