Skip to Content
Merck
All Photos(1)

Key Documents

W384704

Sigma-Aldrich

L-Lysine

≥97%

Synonym(s):

(S)-2,6-Diaminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.19
FEMA Number:
3847
Beilstein:
1722531
EC Number:
MDL number:
UNSPSC Code:
12164502
eCl@ss:
32160406
PubChem Substance ID:
Flavis number:
17.026

biological source

synthetic

Assay

≥97%

optical activity

[α]20/D +24°, c = 2 in 0.6 M HCl

mp

215 °C (dec.) (lit.)

solubility

H2O: 10%, clear, colorless to light greenish-yellow
H2O: 10%, clear, colorless to yellow

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1.0 ppm
Pb: ≤10 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

NCCCC[C@H](N)C(O)=O

InChI

1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)/t5-/m0/s1

InChI key

KDXKERNSBIXSRK-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Faraasen et al.
Pharmaceutical research, 20(2), 237-246 (2003-03-15)
The purpose of this study was to demonstrate specific receptor-mediated targeting of phagocytes by functional surface coatings of microparticles, shielding from nonspecific phagocytosis and allowing ligand-specific interactions via molecular recognition. Coatings of the comb polymer poly(L-lysine)-g-poly(ethylene glycol) (PLL-g-PEG) were investigated
Valentina R Minciacchi et al.
Oncotarget, 6(13), 11327-11341 (2015-04-11)
Large oncosomes (LO) are atypically large (1-10 µm diameter) cancer-derived extracellular vesicles (EVs), originating from the shedding of membrane blebs and associated with advanced disease. We report that 25% of the proteins, identified by a quantitative proteomics analysis, are differentially
Lubin Jiang et al.
Nature, 499(7457), 223-227 (2013-07-05)
The variant antigen Plasmodium falciparum erythrocyte membrane protein 1 (PfEMP1), which is expressed on the surface of P. falciparum-infected red blood cells, is a critical virulence factor for malaria. Each parasite has 60 antigenically distinct var genes that each code
Scott R Floyd et al.
Nature, 498(7453), 246-250 (2013-06-04)
DNA damage activates a signalling network that blocks cell-cycle progression, recruits DNA repair factors and/or triggers senescence or programmed cell death. Alterations in chromatin structure are implicated in the initiation and propagation of the DNA damage response. Here we further
Lunzhi Dai et al.
Nature chemical biology, 10(5), 365-370 (2014-04-01)
We report the identification of a new type of histone mark, lysine 2-hydroxyisobutyrylation (Khib), and identify the mark at 63 human and mouse histone Khib sites, including 27 unique lysine sites that are not known to be modified by lysine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service