Skip to Content
Merck
All Photos(1)

Key Documents

W378917

Sigma-Aldrich

3-(Methylthio)hexyl acetate

≥97%, FG

Synonym(s):

lychee mercaptan acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O2S
CAS Number:
Molecular Weight:
190.30
FEMA Number:
3789
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.236
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

refractive index

n/D 1.4640

bp

216 °C (lit.)

density

0.985 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; vegetable; sulfurous

SMILES string

CCCC(CCOC(C)=O)SC

InChI

1S/C9H18O2S/c1-4-5-9(12-3)6-7-11-8(2)10/h9H,4-7H2,1-3H3

InChI key

VIQXICKUKPVFRK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service