Skip to Content
Merck
All Photos(1)

Key Documents

W369209

Sigma-Aldrich

Hexyl trans-2-hexenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
3692
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.292

biological source

synthetic

SMILES string

CCCCCCOC(=O)\C=C\CCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service