Skip to Content
Merck
All Photos(1)

Key Documents

W367605

Sigma-Aldrich

1-Ethylhexyl tiglate

≥95%, FG

Synonym(s):

3-Octyl tiglate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH(C2H5)(CH2)4CH3
CAS Number:
Molecular Weight:
212.33
FEMA Number:
3676
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.539
NACRES:
NA.21
Organoleptic:
spicy
grade:
FG
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥95%

refractive index

n20/D 1.444 (lit.)

density

0.882 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

spicy

SMILES string

CCCCCC(CC)OC(=O)\C(C)=C\C

InChI

1S/C13H24O2/c1-5-8-9-10-12(7-3)15-13(14)11(4)6-2/h6,12H,5,7-10H2,1-4H3/b11-6+

InChI key

LMBAQNNBRWRROG-IZZDOVSWSA-N

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service