Skip to Content
Merck
All Photos(1)

Documents

W351200

Sigma-Aldrich

2-Methyltetrahydrothiophen-3-one

≥97%, FG

Synonym(s):

blackberry thiophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8OS
CAS Number:
Molecular Weight:
116.18
FEMA Number:
3512
EC Number:
Council of Europe no.:
11601
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.023
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.508 (lit.)

bp

82 °C/28 mmHg (lit.)

density

1.119 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; fruity; sulfurous

SMILES string

CC1SCCC1=O

InChI

1S/C5H8OS/c1-4-5(6)2-3-7-4/h4H,2-3H2,1H3

InChI key

YMZZPMVKABUEBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service