Skip to Content
Merck
All Photos(1)

Documents

W349100

Sigma-Aldrich

2-Ethylfenchol

≥97%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O
CAS Number:
Molecular Weight:
182.30
FEMA Number:
3491
Beilstein:
3044187
EC Number:
Council of Europe no.:
10208
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.095
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC

Assay

≥97%

optical activity

[α]20/D −12°, c = 1.5 in ethanol

refractive index

n20/D 1.482 (lit.)

bp

105 °C/15 mmHg (lit.)

density

0.956 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

earthy

SMILES string

CCC1(O)C(C)(C)[C@@H]2CC[C@@]1(C)C2

InChI

1S/C12H22O/c1-5-12(13)10(2,3)9-6-7-11(12,4)8-9/h9,13H,5-8H2,1-4H3/t9-,11+,12?/m1/s1

InChI key

KIPCKEJKGCXRGA-BVAQLPTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylfenchol is a tertiary alcohol that produces an earthy odor similar to geosmin.

Biochem/physiol Actions

Taste at 10 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tertiary alcohols with earthy odor.
Napolitano E, et al.
Journal of Agricultural and Food Chemistry, 44(9), 2806-2809 (1996)
Odor similarities in structurally related odorants.
Polak E, et al.
Chemical Senses, 3(4), 369-380 (1978)
Synthesis of new earthy odorants.
Finato B, et al.
Journal of Agricultural and Food Chemistry, 40(5), 857-859 (1992)
Mosciano, G.
Perfumer & Flavorist, 30, 57-57 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 41-41 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service