Skip to Content
Merck
All Photos(1)

Key Documents

W309101

Sigma-Aldrich

γ-Undecalactone

natural (US), ≥98%, FG

Synonym(s):

Undecanoic γ-lactone, Aldehyde C14

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3091
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.002
NACRES:
NA.21

grade

FG
Halal
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥98%

refractive index

n20/D 1.45 (lit.)

bp

164-166 °C/13 mmHg (lit.)

density

0.949 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; creamy; fatty; fruity; peach

SMILES string

CCCCCCCC1CCC(=O)O1

InChI

1S/C11H20O2/c1-2-3-4-5-6-7-10-8-9-11(12)13-10/h10H,2-9H2,1H3

InChI key

PHXATPHONSXBIL-UHFFFAOYSA-N

Legal Information

For R&D Use and/or Food Manufacturing only

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service