Skip to Content
Merck
All Photos(1)

Documents

W295101

Sigma-Aldrich

Propyl 4-hydroxybenzoate

≥99%

Synonym(s):

4-Hydroxybenzoic acid propyl ester, Propyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH2CH2CH3
CAS Number:
Molecular Weight:
180.20
FEMA Number:
2951
Beilstein:
1103245
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

grade

Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥99%

mp

95-98 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

burnt; smoky

SMILES string

CCCOC(=O)c1ccc(O)cc1

InChI

1S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11H,2,7H2,1H3

InChI key

QELSKZZBTMNZEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propyl 4-hydroxybenzoate belongs to a group called parabens, which are commonly used as preservatives in cosmetics, skin care products, food, and beverages. Propyl 4-hydroxybenzoate, in combination with other parabens also shows strong antimicrobial activity against a wide range of micro-organisms.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

356.0 °F

Flash Point(C)

180 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of HPLC analysis of 2-phenoxyethanol, 1-phenoxypropan-2-ol, methyl, ethyl, propyl, butyl and benzyl 4-hydroxybenzoate (parabens) in cosmetic products, with emphasis on decision limit and detection capability.
Borremans M, et al.
Chromatographia, 59(1), 47-53 (2004)
Dennis G Maki et al.
Critical care medicine, 39(4), 613-620 (2011-01-05)
Catheter-related bloodstream infection is the greatest threat to the safety of patients on hemodialysis. Catheter lock solutions containing heparin have been linked to an increased risk of hemorrhage and thrombocytopenia. To ascertain the safety and efficacy for prevention of catheter-related
Miguel González-Doncel et al.
Environmental pollution (Barking, Essex : 1987), 184, 360-369 (2013-10-08)
Here we proposed a battery of non-invasive biomarkers and a histological survey to examine physiological/anatomical features in embryos, eleutheroembryos (13 days post-fertilization, dpf), and larvae (28-42 dpf) of medaka to investigate the effects of embryonic exposure to propylparaben (PrP). Concentrations <1000 μg
C L Barberis et al.
Journal of food protection, 73(8), 1493-1501 (2010-09-08)
Each year, a significant portion of the peanuts produced cannot be marketed because of fungal disease at the postharvest stage and mycotoxin contamination. Antioxidants could be used as an alternative to fungicides to control ochratoxigenic fungi in peanuts during storage.
Yang Wang et al.
Analytica chimica acta, 673(2), 145-150 (2010-07-06)
A selective, sensitive, rapid and reliable method based on molecularly imprinted polymers (MIPs) with dual templates to determine total content of parabens in cosmetics was developed. With methylparaben (MP) and propylparaben (PP) as dual-templates, methacrylic acid (MAA) as a functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service