Skip to Content
Merck
All Photos(2)

Documents

W291113

Sigma-Aldrich

Piperonal

natural, 98%, FG

Synonym(s):

1,3-Benzodioxole-5-carboxaldehyde, 3,4-(Methylenedioxy)benzaldehyde, Heliotropin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
FEMA Number:
2911
Beilstein:
131691
EC Number:
Council of Europe no.:
104
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.016

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 182.60

vapor pressure

1 mmHg ( 87 °C)

Assay

98%

composition

contains EU 1223/2009 restricted and IFRA restricted Safrole

bp

264 °C (lit.)

mp

35-39 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

heliotropine

Organoleptic

coconut; floral; sweet; vanilla

SMILES string

[H]C(=O)c1ccc2OCOc2c1

InChI

1S/C8H6O3/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-4H,5H2

InChI key

SATCULPHIDQDRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Natural occurrence: Tahitian vanilla, camphor wood, violet flowers, burley tobacco, dill and sherry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

150.1 °F

Flash Point(C)

65.62 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Peock et al.
Journal of the Royal Society of Health, 113(6), 292-294 (1993-12-01)
Piperonal, once used to kill lice in Australian hospitals, was acclaimed as an effective pediculicide (Corlette, 1925) by the standards of the day. It is unusual in also exhibiting a repellent action against lice, a property only recently realised. A
Ariel Ceferino Toloza et al.
Memorias do Instituto Oswaldo Cruz, 101(1), 55-56 (2006-05-16)
New alternative insecticides are necessary for the chemical control of head lice. In this study the fumigant knockdown time 50% (KT50) and repellency index (RI) of three aliphatic lactones was compared with two essential oils and DDVP, against permethrin-resistance Pediculus
A R Dahl et al.
Biochemical pharmacology, 34(5), 631-636 (1985-03-01)
Eighteen methylenedioxyphenyl (MDP) compounds, including some commonly inhaled by people, were tested for the ability to inhibit rabbit nasal microsomal cytochrome P-450-dependent hexamethylphosphoramide (HMPA) N-demethylase. For comparison, liver microsomes were also used. Nasal cytochrome P-450 from rabbits metabolized MDP compounds
The inhibition of rat nasal cytochrome P-450-dependent mono-oxygenase by the essence heliotropin (piperonal).
A R Dahl
Drug metabolism and disposition: the biological fate of chemicals, 10(5), 553-554 (1982-09-01)
Michael S Macias et al.
Forensic science international, 195(1-3), 132-138 (2010-01-02)
Currently, in the field of odor detection, there is generally a wider variation in limit of detections (LODs) for canines than instruments. The study presented in this paper introduces an improved protocol for the creation of controlled odor mimic permeation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service