Skip to Content
Merck
All Photos(1)

Key Documents

W283823

Sigma-Aldrich

Patchouli oil

Certified organic (NOP)

Synonym(s):

Pogostemon cablin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2838
MDL number:
UNSPSC Code:
12164502

biological source

Pogostemon cablin (Blanco) Benth

grade

Certified organic (NOP)
Halal
Kosher

refractive index

n20/D 1.509 (lit.)

bp

287 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

earthy

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service