Skip to Content
Merck
All Photos(1)

Key Documents

W264601

Sigma-Aldrich

Linalyl isovalerate

≥95%, FG

Synonym(s):

Linalyl 3-methylbutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
238.37
FEMA Number:
2646
EC Number:
Council of Europe no.:
449
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.454
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.451 (lit.)

bp

188 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; sage; sweet

SMILES string

CC(C)CC(=O)OC(C)(CC\C=C(\C)C)C=C

InChI

1S/C15H26O2/c1-7-15(6,10-8-9-12(2)3)17-14(16)11-13(4)5/h7,9,13H,1,8,10-11H2,2-6H3

InChI key

WCDGWAIZRYMVOW-UHFFFAOYSA-N

General description

Linalyl isovalerate is commonly used as a fragrance ingredient.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RIFM fragrance ingredient safety assessment, Linalyl isovalerate, CAS Registry Number 1118-27-0.
Api AM, et al.
Food And Chemical Toxicology, 84, S88-S99 (2015)
C S Letizia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(7), 1011-1015 (2003-06-14)
A toxicologic and dermatologic review of linalyl isovalerate when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service