Skip to Content
Merck
All Photos(1)

Key Documents

W254304

Sigma-Aldrich

2,3-Heptanedione

≥97%, FG

Synonym(s):

Acetyl valeryl

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
R$353.00
W254304-SAMPLE-K
R$373.00
1 KG
R$4,583.00

R$353.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
100 G
R$353.00
W254304-SAMPLE-K
R$373.00
1 KG
R$4,583.00

About This Item

Linear Formula:
CH3(CH2)3COCOCH3
CAS Number:
Molecular Weight:
128.17
FEMA Number:
2543
EC Number:
Council of Europe no.:
2044
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.064
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

R$353.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.415 (lit.)

bp

64 °C/18 mmHg (lit.)

density

0.92 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

butter; cheese; oily

SMILES string

CCCCC(=O)C(C)=O

InChI

1S/C7H12O2/c1-3-4-5-7(9)6(2)8/h3-5H2,1-2H3

InChI key

FJPGAMCQJNLTJC-UHFFFAOYSA-N

Gene Information

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edyta Budzyńska et al.
Talanta, 209, 120594-120594 (2020-01-02)
Electronic cigarettes (e-cigarettes) are continuously increasing in popularity due to being considered healthier compared with traditional cigarettes. Based on the European directive 2014/40/EU [1] the German legislation restricts the usage of potential harmful substances in these tobacco related products [2,3].
Jan Hintzpeter et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(1), 263-273 (2014-11-08)
The purpose of this study was to investigate the origin and function of the aldo-keto reductase (AKR) superfamily as enzymes involved in the detoxification of xenobiotics. We used the cyanobacterium Synechocystis sp. PCC 6803 as a model organism and sequence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service