Skip to Content
Merck
All Photos(3)

Key Documents

W242810

Sigma-Aldrich

Ethyl isobutyrate

greener alternative

natural, ≥98%, FCC, FG

Synonym(s):

2-methyl propionic acid ethyl ester, Ethyl 2-methylpropanoate, Ethyl isobutanoate

Sign Into View Organizational & Contract Pricing

Select a Size

W242810-SAMPLE-K
R$347.00
100 G
R$482.00
1 KG
R$4,470.00

R$347.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W242810-SAMPLE-K
R$347.00
100 G
R$482.00
1 KG
R$4,470.00

About This Item

Linear Formula:
(CH3)2CHCOOC2H5
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2428
Beilstein:
773846
EC Number:
Council of Europe no.:
288c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.413
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

R$347.00


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

4.01 (vs air)

vapor pressure

40 mmHg ( 33.8 °C)

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.387 (lit.)

bp

112-113 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

fruity; ethereal; sweet

SMILES string

CCOC(=O)C(C)C

InChI

1S/C6H12O2/c1-4-8-6(7)5(2)3/h5H,4H2,1-3H3

InChI key

WDAXFOBOLVPGLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philippe Renault et al.
International journal of food microbiology, 207, 40-48 (2015-05-24)
The aim of this work was to study ester formation and the aromatic impact of Torulaspora delbrueckii when used in association with Saccharomyces cerevisiae during the alcoholic fermentation of must. In order to evaluate the influence of the inoculation procedure
Georgia Lytra et al.
Food chemistry, 194, 196-200 (2015-10-17)
Our study focused on variations in wine aroma perception and molecular composition during tasting over a period of 30min. In parallel, dynamic analytical and sensory methods were applied to study changes in the wines' molecular and aromatic evolution. Dynamic sensory
Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not
Stéphanie Rollero et al.
Applied microbiology and biotechnology, 99(5), 2291-2304 (2014-11-22)
Volatile compounds produced by yeast during fermentation greatly influence the organoleptic qualities of wine. We developed a model to predict the combined effects of initial nitrogen and phytosterol content and fermentation temperature on the production of volatile compounds. We used
Pascal Fuchsmann et al.
Journal of agricultural and food chemistry, 63(34), 7511-7521 (2015-08-01)
To establish the odor profiles of three differently fabricated commercial Swiss Tilsit cheeses, analyses were conducted using headspace solid-phase microextraction gas chromatography-mass spectrometry/pulsed flame photometric detection and gas chromatography-olfactometry to identify and quantitate volatile compounds. In addition, odor quality and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service