Skip to Content
Merck
All Photos(1)

Key Documents

W236802

Sigma-Aldrich

Decyl butyrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2(CH2)9CH3
CAS Number:
Molecular Weight:
228.37
FEMA Number:
2368
EC Number:
Council of Europe no.:
273
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.047

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

Assay

97%

refractive index

n20/D 1.431 (lit.)

bp

134-135 °C/8 mmHg (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; citrus; waxy

SMILES string

CCCCCCCCCCOC(=O)CCC

InChI

1S/C14H28O2/c1-3-5-6-7-8-9-10-11-13-16-14(15)12-4-2/h3-13H2,1-2H3

InChI key

PUCQHFICPFUPKW-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service