Skip to Content
Merck
All Photos(1)

Key Documents

W236306

Sigma-Aldrich

Decanal dimethyl acetal

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$28.75
250 MG
$50.60
1 G
$127.65
5 G
$447.35

Vendor SKU: A139036-100MG

$28.75


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$28.75
250 MG
$50.60
1 G
$127.65
5 G
$447.35

About This Item

Linear Formula:
CH3(CH2)8CH(OCH3)2
CAS Number:
Molecular Weight:
202.33
FEMA Number:
2363
EC Number:
Council of Europe no.:
43
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.009

Vendor SKU: A139036-100MG

$28.75


Ships in 7 days - from Aldrich Partner.

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.424 (lit.)

bp

218 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service