Skip to Content
Merck
All Photos(1)

Key Documents

W235900

Sigma-Aldrich

Davana oil

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2359
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Kosher
natural
biological source:
Artemisia pallens
food allergen:
no known allergens

biological source

Artemisia pallens

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

form

liquid

composition

Limonene, 0.5%

refractive index

n20/D 1.488

density

0.958 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; sweet

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service