Skip to Content
Merck
All Photos(1)

Key Documents

W234818

Sigma-Aldrich

Cyclohexaneethyl acetate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH2C6H11
CAS Number:
Molecular Weight:
170.25
FEMA Number:
2348
EC Number:
Council of Europe no.:
218
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.028

Assay

≥98%

refractive index

n20/D 1.448 (lit.)

bp

222 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

CC(=O)OCCC1CCCCC1

InChI

1S/C10H18O2/c1-9(11)12-8-7-10-5-3-2-4-6-10/h10H,2-8H2,1H3

InChI key

NOTFZGFABLVTIG-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service