Skip to Content
Merck
All Photos(1)

Documents

W233100

Sigma-Aldrich

Cognac oil

green, natural, FG

Synonym(s):

Vitis vinifera, Grape seed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2331
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Vitis vinifera

grade

FG
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.50

form

liquid

refractive index

n20/D 1.428

density

0.870 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; wine-like

Looking for similar products? Visit Product Comparison Guide

General description

Cognac oil is mainly found in wine lees, produced due to the fermentation of yeast and other sediments. It is responsible for the cognac aroma of the wine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.1 °F - closed cup

Flash Point(C)

66.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenaroli's Handbook of Flavor Ingredients, Fourth Edition, 341-341 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service