Skip to Content
Merck
All Photos(1)

Key Documents

W231126

Sigma-Aldrich

Citronellyl acetate

natural

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
2311
Council of Europe no.:
202c
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.012

grade

natural

refractive index

n20/D 1.445 (lit.)

bp

240 °C (lit.)

density

0.891 g/mL at 25 °C (lit.)

SMILES string

CC(CCOC(C)=O)CC\C=C(\C)C

InChI

1S/C12H22O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,11H,5,7-9H2,1-4H3

InChI key

JOZKFWLRHCDGJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Förster-Fromme et al.
Microbiology (Reading, England), 154(Pt 3), 789-796 (2008-03-04)
The atuRABCDEFGH gene cluster is essential for acyclic terpene utilization (Atu) in Pseudomonas aeruginosa. The biochemical functions of most Atu proteins have not been experimentally verified; exceptions are AtuC/AtuF, which constitute the two subunits of geranyl-CoA carboxylase, the key enzyme
S Oda et al.
Bioscience, biotechnology, and biochemistry, 65(8), 1917-1919 (2001-10-02)
A double coupling system, which couples metabolism of glucose and transacetylation, is a unique procedure for the production of acetic esters. In the novel coupling system described in this article, acetyl coenzyme A (acetyl-CoA) was supplied via metabolism of both
Hyang-Sook Choi
Journal of agricultural and food chemistry, 53(5), 1642-1647 (2005-03-03)
This study was conducted to determine the composition of kumquat (Fortunella japonica Swingle) cold-pressed peel oil and to determine which volatile components are primarily responsible for the aroma of this oil. Eighty-two compounds were identified in the oil by GC
H J Fang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 24(5), 366-371 (1989-01-01)
Three geranium oils were separated, identified and compared by high resolution cross-linked fused silica capillary GC and GC-MSD. It was found that their chemical constituents are very similar, but the contents of some components (linalool, citronellol, cis-caryophyllene and guaiene) differ
Jun Han et al.
Pest management science, 67(12), 1583-1588 (2011-06-16)
This study was aimed at assessing the fumigant toxicity of 14 essential oil constituents from lemon eucalyptus, Eucalyptus citriodora Hook, and another ten known compounds to females of acaricide-susceptible, chlorfenapyr-resistant, fenpropathrin-resistant, pyridaben-resistant and abamectin-resistant strains of Tetranychus urticae Koch. Menthol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service