Recommended Products
biological source
synthetic
Quality Level
grade
Kosher
reg. compliance
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515
Assay
≥92%
refractive index
n20/D 1.553 (lit.)
bp
250-254 °C (lit.)
density
1.08 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
balsam; green
SMILES string
O=COC\C=C\c1ccccc1
InChI
1S/C10H10O2/c11-9-12-8-4-7-10-5-2-1-3-6-10/h1-7,9H,8H2/b7-4+
InChI key
LBHJXKYRYCUGPD-QPJJXVBHSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service