Skip to Content
Merck
All Photos(1)

Documents

W229301

Sigma-Aldrich

Cinnamyl acetate

natural, 95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=CHC6H5
CAS Number:
Molecular Weight:
176.21
FEMA Number:
2293
EC Number:
Council of Europe no.:
208
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.018
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

95%

refractive index

n20/D 1.541 (lit.)

bp

265 °C (lit.)

density

1.052 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cinnamon; floral; spicy; sweet

SMILES string

CC(=O)OC\C=C\c1ccccc1

InChI

1S/C11H12O2/c1-10(12)13-9-5-8-11-6-3-2-4-7-11/h2-8H,9H2,1H3/b8-5+

InChI key

WJSDHUCWMSHDCR-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nehad E M Taktak et al.
Pesticide biochemistry and physiology, 158, 156-165 (2019-08-06)
Culex pipiens is a main vector for Bancroftian filariasis, Rift Valley Fever and diseases caused by other viruses, leaving several peoples with disabilities. In recent years, plant derived compounds have received much attention as potential alternatives to synthetic chemicals due
Emilie Deletre et al.
Parasites & vectors, 8, 316-316 (2015-06-13)
Laboratory and field studies showed that repellent, irritant and toxic actions of common public health insecticides reduce human-vector contact and thereby interrupt disease transmission. One of the more effective strategies to reduce disease risk involves the use of long-lasting treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service