Skip to Content
Merck
All Photos(1)

Key Documents

W207500

Sigma-Aldrich

Isoamyl hexanoate, mixture of isomers

FCC

Synonym(s):

Isoamyl caproate

Sign Into View Organizational & Contract Pricing

Select a Size

W207500-SAMPLE-K
R$373.00
1 KG
R$526.00
9 KG
R$4,238.00

R$373.00


Estimated to ship onApril 07, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W207500-SAMPLE-K
R$373.00
1 KG
R$526.00
9 KG
R$4,238.00

About This Item

Empirical Formula (Hill Notation):
C11H22O2
CAS Number:
Molecular Weight:
186.29
FEMA Number:
2075
Council of Europe no.:
320
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.070
NACRES:
NA.21
Organoleptic:
apple; green; fruity; pineapple; sweet
grade:
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

R$373.00


Estimated to ship onApril 07, 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

98%

refractive index

n20/D 1.42 (lit.)

bp

222 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green; fruity; pineapple; sweet

SMILES string

CCCCCC(=O)OCCC(C)C

InChI

1S/C11H22O2/c1-4-5-6-7-11(12)13-9-8-10(2)3/h10H,4-9H2,1-3H3

InChI key

XVSZRAWFCDHCBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service