Documents
THS01115
ThalesNano CatCart® catalyst cartridge system, 30 mm L
20%Pd(OH)2/C
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
UNSPSC Code:
60104500
NACRES:
NB.51
packaging
pack of 6 ea
manufacturer/tradename
ThalesNano THS 01115
diam.
4 mm
Related Categories
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.