Key Documents
TEM-LC400-NI-100
Sigma-Aldrich
Lacey Carbon Supported Nickel Grid
size 400 mesh, hole size 100 μm , Average, box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-LC400-NI-100 is currently not available for purchase. Contact Technical Service
packaging
box of 25
hole size
100 μm , Average
size
400 mesh
General description
Lacey Carbon Supported Grid: Nickel Grid, 400 mesh. Average hole size is 100 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)