Documents
TEM-CF200F2CU
Sigma-Aldrich
Carbon Film Supported Copper Gilder Finder F2 Grids
size 200 mesh, box of 25
Synonym(s):
CARBON FILM ONLY ON GILDER FI FINDER 200# CU F2 FINDER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported glider finder (F2) grids. Copper grid, 200 mesh. Gilder finder grids are locator grids with reference/index on the grid: 3.05mm diameter, 0.7 mil (18μm) thickness. F2 type of design has thick bars dividing regions into 9 thin bar areas, which are identified by alphabetical letters located in the center of the grid.