Skip to Content
Merck
All Photos(1)

Key Documents

T9627

Sigma-Aldrich

2-Tetradecanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CH(OH)CH3
CAS Number:
Molecular Weight:
214.39
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

35-36 °C (lit.)

SMILES string

CCCCCCCCCCCCC(C)O

InChI

1S/C14H30O/c1-3-4-5-6-7-8-9-10-11-12-13-14(2)15/h14-15H,3-13H2,1-2H3

InChI key

BRGJIIMZXMWMCC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumikazu Takahashi et al.
FEMS yeast research, 19(2) (2019-02-13)
The yeast Starmerella bombicola NBRC10243 is an excellent producer of sophorolipids, which are among the most useful biosurfactants. The primary alcoholic metabolic pathway of S. bombicola has been elucidated using alcohol oxidase FAO1, but the secondary alcohol metabolic pathway remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service