Skip to Content
Merck
All Photos(2)

Key Documents

T12807

Sigma-Aldrich

2(3)-(Tetrahydrofurfuryloxy)tetrahydropyran

98%

Synonym(s):

‘T-Solvent’

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
R$2,373.00

R$2,373.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
R$2,373.00

About This Item

Empirical Formula (Hill Notation):
C10H18O3
CAS Number:
Molecular Weight:
186.25
Beilstein:
109779
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$2,373.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.461 (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

C1CCC(OC1)OCC2CCCO2.C3COCC(C3)OCC4CCCO4

InChI

1S/2C10H18O3/c1-3-9(7-11-5-1)13-8-10-4-2-6-12-10;1-2-6-12-10(5-1)13-8-9-4-3-7-11-9/h2*9-10H,1-8H2

InChI key

OMNURHQDSBSXKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service