Documents
SYNS321230
Synthware straight bore glass stopcock
bore size 2 mm, O.D. 8 mm stem, plug size 12/30
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S321230
bore size
2 mm
Related Categories
General description
Plugs and barrels are ground to be interchangeable, with 1:10 taper.
Legal Information
Synthware is a trademark of Kemtech America Inc.