Key Documents
SYNS312044
Sigma-Aldrich
Synthware T-bore glass stopcock Entire assembly
bore size 6 mm, plug size 20/44
Synonym(s):
Synthware S312044, t-shape stopcock
Select a Size
1 EA
R$347.00
R$347.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$347.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
R$347.00
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S312044
bore size
6 mm
Related Categories
General description
With solid glass plug. With constant pressure retention spring and clip.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)