Key Documents
SYNM300005
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 5, Hose Connections: Front-right, Rear-left-right
Select a Size
1 EA
R$3,382.00
R$3,382.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$3,382.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
R$3,382.00
Check Cart for Availability
feature
number of ports 5
packaging
pack of 1 ea
manufacturer/tradename
M300005
General description
Synthware glass manifold featuring 5 ports, high vacuum solid glass stopcocks that are precision ground with corresponding numbers on the barrel and plug. The solid glass stopcocks have constant pressure retention devices. The beams have a 22 mm O.D. with front-right and rear-left 10 mm hose connections. The distance between the stopcocks is 75 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)