Key Documents
SYNF794250
Synthware modified evaporation flask
neck joint: ST/NS 24/40, capacity 250 mL
Synonym(s):
Synthware F794250, rotary evaporator flask
Select a Size
1 EA
R$191.00
R$191.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$191.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
R$191.00
Check Cart for Availability
material
glass flask
round bottom flask
Joint
joint: ST/NS neck24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F794250
capacity
250 mL
General description
Low profile design sits upright on bench and also provides for easy product access with a spatula. The unique flask shape is excellent for the controlled evaporation of foaming solvents such as toluene or xylene on a rotary evaporator
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)