Key Documents
SYNF641125
Synthware pressure equalizing funnel with high vacuum valve
125 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(s):
Synthware F671425, dropping funnel
Select a Size
1 EA
R$672.00
R$672.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$672.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
R$672.00
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F641125
capacity
125 mL
125 mL
Related Categories
General description
With PTFE extended tip valve for semi-needle valve adjustment.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)