Key Documents
SYNF490500
Synthware storage flask with high vacuum valve and side arm
500 mL, valve size 0-4 mm
Synonym(s):
Synthware F490500, round bottom flask
Select a Size
1 EA
R$401.00
R$401.00
Available to ship onMarch 06, 2025Details
Select a Size
Change View
1 EA
R$401.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
R$401.00
Available to ship onMarch 06, 2025Details
material
borosilicate glass flask
round bottom
single-neck (high vacuum valve)
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F490500
capacity
500 mL
500 mL
valve size
0-4 mm
General description
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper( “umbrella”) to protect Viton® O-ring on shaft..
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)