Skip to Content
Merck
All Photos(1)

Documents

SYNF392475

Synthware powder funnel with 60 degree offset

joint: ST/NS 24/40, top diam. 75 mm

Synonym(s):

Synthware F392475, glass powder funnel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.51

Joint

joint: ST/NS24/40

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F392475

top diam.

75 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For use with multi-neck flasks.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service