Key Documents
SYNF339100
Synthware single-neck round-bottom flask with joint hooks
100 mL, joint: ST/NS 19/22
Synonym(s):
Synthware F339100, flask with hooks
Select a Size
1 EA
R$108.00
R$108.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$108.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
R$108.00
Check Cart for Availability
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F339100
capacity
100 mL
100 mL
General description
Heavy wall design.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)