Key Documents
SYNF221500
Synthware four neck round bottom flask with angled side necks
capacity 500 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(s):
Synthware F221500, 4-neck reaction flask
Select a Size
1 EA
R$432.00
R$432.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$432.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
R$432.00
Check Cart for Availability
material
borosilicate glass flask
four-neck
round bottom
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F221500
capacity
500 mL
General description
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)