SYNF192975
Synthware™ powder funnel with flattened side
joint: ST/NS 29/42, top diam. 75 mm
Synonym(s):
Synthware F192975, glass powder funnel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Joint
joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F192975
top diam.
75 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Funnels have 60° angled side with a standard taper inner joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service