Key Documents
SYNC554300
Synthware West condenser with removable hose connections
joint: ST/NS 24/40, jacket L 300 mm
Synonym(s):
Synthware C554300, glass West condenser
Select a Size
1 EA
R$363.00
R$363.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$363.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
R$363.00
Check Cart for Availability
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C554300
jacket L
300 mm
Related Categories
General description
With narrow annular space for high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)