Key Documents
SYNA971922
Synthware distilling adapter with compression cap
joint: ST/NS 19/22, side arm angle: 75 deg
Synonym(s):
Synthware A971922, adapter, distillation adapter
Select a Size
1 EA
R$182.00
R$182.00
Estimated to ship onMay 30, 2025
Select a Size
Change View
1 EA
R$182.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
R$182.00
Estimated to ship onMay 30, 2025
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A971922
General description
With compression cap (RC009) and Viton® O-ring for forming a vacuum tight seal of plain thermometer.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)