Key Documents
SYNA871922
Synthware 75 degree angled drying tube
joint: ST/NS 19/22
Select a Size
1 EA
R$126.00
R$126.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$126.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
R$126.00
Check Cart for Availability
material
colorless tube
glass tube
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A871922
General description
Angled at 75° to avoid desiccant contact with reactions.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)