Key Documents
SYNA663514
Synthware adapter with top spherical socket joint and lower standard taper inner joint
top joint: ST/NS 35/20, bottom joint: ST/NS 14/20
Synonym(s):
Synthware A663514, adapter with ball socket
Select a Size
1 EA
R$136.00
R$136.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$136.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
R$136.00
Check Cart for Availability
Joint
bottom joint: ST/NS14/20
joint: ST/NS top35/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A663514
Related Categories
General description
Synthware adapter with top spherical socket joint (35/20) and lower standard taper inner joint (14/20) to fit a various of laboratory glassware.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)