SYNA551922
Synthware™ distillation distribution adapter
joint: ST/NS 19/22
Synonym(s):
Synthware A551922, cow distillation adapter
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A551922
Related Categories
General description
Rotating cow receiver allows four flasks to be progressively filled.
Other Notes
includes only the top part
Legal Information
Synthware is a trademark of Kemtech America Inc.