Documents
SYNA502440
Synthware gas inlet adapter
joint: ST/NS 24/40, O.D. × L 153 mm
Synonym(s):
Synthware A502440, gas inlet adapter, extended drip tube
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A502440
O.D. × L
153 mm
Related Categories
General description
Inner joint with a sealed on lower tube extending 6” below the grind, having the same O.D. tubing on both ends of the grind.
Legal Information
Synthware is a trademark of Kemtech America Inc.