Key Documents
SYNA111420
Synthware modified Claisen adapter
joint: ST/NS 14/20
Synonym(s):
Synthware A111420, distilling adapter
Select a Size
1 EA
R$324.00
R$324.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$324.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
R$324.00
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A111420
Related Categories
General description
Three-inlet type for multiple entries into a single neck flask. One side joint is at a 45° angle.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)