SM601001010
SmartPor, alumina membrane
thickness 100 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Pore geometry: trigonal arrangement
packaging
pkg of 10 pieces
size
10 mm × 10 mm
thickness
100 μm
pore size
125 nm (interpore distance)
60 nm pore diameter
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service