Skip to Content
Merck
All Photos(1)

Documents

SLW4526/45D

Pyrex® High temperature screwcap for Media-lab bottle

red, thread suits 45 mm

Synonym(s):

Pyrex, SciLabware 4526/45D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

PTFE liner
polyester cap
red

manufacturer/tradename

4526/45D
SciLabware

fitting

thread suits 45 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • For use with 1516, 1517, 1518, 1519 and 1520 series bottles
  • Ideal for applications involving high temperatures or aggressive chemicals
  • Red thermoplastic polyester cap with natural coloured PTFE faced sealing disc
  • Withstands hot air sterilisation up to 180 deg. C
  • PTFE sealing disc resists chemical attack

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service