Skip to Content
Merck
All Photos(1)

Documents

SLW3215/10P

Pyrex® Mixing cylinder, class B, detachable foot,

100 mL

Synonym(s):

Pyrex, SciLabware 3215/10P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

polyethylene base
polyethylene stopper

Joint

joint stopper24/29

feature

graduations 1.0 mL
detachable foot

manufacturer/tradename

3215/10P
SciLabware

H

270 mm

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Complies with ISO 4788
  • Comprises borosilicate glass cylinder, polyethylene foot, and polyethylene stopper
  • Replacement parts available separately
  • Graduations in white enamel

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service