SLW1519/06
Pyrex® Media-Lab Bottles, amber glass, with-out cap and pouring ring, with printed trace code
capacity 250 mL
Synonym(s):
Pyrex, SciLabware 1519/06
All Photos(1)
About This Item
Recommended Products
material
round amber glass bottle
Joint
Joints threaded neck (GL45)
feature
graduations
without cap
with printed trace code
without pouring ring
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1519/06
H
138 mm
body diam.
70 mm
capacity
250 mL
General description
- Bottles only. Supplied without cap or pouring ring
- Complies with ISO 4796
- Manufactured from amber-coated borosilicate glass
- UV absorbent up to 500 nm
- Ideal for storage of light sensitive materials
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
Features and Benefits
• Bottles only. Supplied without cap or pouring ring• Complies with ISO 4796• Manufactured from amber-coated borosilicate glass• UV absorbent up to 500 nm• Ideal for storage of light sensitive materials• Printed with Trace Code for downloadable batch certificate• The 25 ml size bottle features a glass lip and does not require a pouring ring.• Never autoclave bottles with a fully tightened screw cap. Always slacken off. Failure to follow this procedure can lead to breakage of the bottle.
Legal Information
PYREX is a registered trademark of Corning, Inc.